Anti-PDZK1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The PDZK1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PDZK1. This antibody reacts with human, mouse, bovine, rat (negative). The PDZK1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400
Type: Primary
Antigen: PDZK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Bovine, Rat (Negative)
Specifications
- Catalog No:
- NOVUNB400-149SS
- NOVUNB400-149
- Antigen symbol:
- PDZK1
- PDZK1
- Antigen name:
- PDZ Domain Containing 1
- PDZ Domain Containing 1
- Antigen synonyms:
- NHERF-3|NHERF3|CLAMP|CAP70|PDZD1
- NHERF-3|NHERF3|CLAMP|CAP70|PDZD1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 5174
- 5174
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice