Anti-ABCG8 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: ABCG8 (ATP-binding cassette, sub-family G (WHITE), member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB400-110
- NOVUNB400-110SS
- Antigen symbol:
- ABCG8
- ABCG8
- Antigen name:
- ATP Binding Cassette, sub-family G, member 8
- ATP Binding Cassette, sub-family G, member 8
- Antigen synonyms:
- STSL|ATP-binding cassette sub-family G member 8|Sterolin-2|ABCG8|GBD4
- STSL|ATP-binding cassette sub-family G member 8|Sterolin-2|ABCG8|GBD4
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 64241
- 64241
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together