Anti-ABCA1 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB400-105G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family A, member 1
- Antigen symbol:ABCA1
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:19
- Antigen synonyms:ABC-1|ATP-binding cassette transporter 1|CERP|HDLDT1|TGD|ABC1|membrane-bound|ATP-binding cassette transporter A1|cholesterol efflux regulatory protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
The ABCA1 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to ABCA1. This antibody reacts with human, mouse, rat, porcine, canine, chicken, hamster, mustelid. The ABCA1 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: ABCA1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Canine, Chicken, Hamster, Mustelid