Order Entry
Sweden
ContactUsLinkComponent
 
Anti-OGT Mouse Monoclonal Antibody [clone: HGAC85]
 
undefined
Anti-OGT Mouse Monoclonal Antibody [clone: HGAC85]

 

  • Catalog No:
  • NOVUNB300-614SS
  • NOVUNB300-614
  • Antigen symbol:
  • OGT
  • O-GlcNAc
  • Antigen name:
  • O-linked N-acetylglucosamine (GlcNAc) transferase
  • O-linked N-acetylglucosamine
  • Antigen synonyms:
  • O-GlcNAc transferase subunit p110|O-linked N-acetylglucosamine transferase 110 kDa subunit|UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit|OGT
  • O-GlcNAc transferase p110 subunit|O-GlcNAc transferase subunit p110|O-linked N-acetylglucosamine transferase 110 kDa subunit|O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase)|O-linked GlcNAc transferase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • HGAC85
  • HGAC85
  • Reactivity:
  • Multiple
  • Multiple
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG3 kappa
  • IgG3 kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • ChIP:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice