Order Entry
Sweden
ContactUsLinkComponent
 
Anti-CDKN2A Rabbit Polyclonal Antibody
 
undefined
Anti-CDKN2A Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNB200-106SS
  • NOVUNB200-106
  • Antigen symbol:
  • p19ARF
  • p19ARF
  • Antigen name:
  • Cyclin-dependent kinase inhibitor 2A
  • Cyclin-dependent kinase inhibitor 2A
  • Antigen synonyms:
  • CDKN2|p16-INK4|MLM|INK4|P16|p19ARF|p16-INK4a|p16INK4a|p16|CDKN2A|p16INK4|p19|p14|p16INK4A|CDK4I|INK4a|p14ARF|CMM2|ARF|MTS1|TP16
  • CDKN2|p16-INK4|MLM|INK4|P16|p19ARF|p16-INK4a|p16INK4a|p16|CDKN2A|p16INK4|p19|p14|p16INK4A|CDK4I|INK4a|p14ARF|CMM2|ARF|MTS1|TP16
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 12578
  • 12578
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice