Anti-Pyruvate Dehydrogenase E1-alpha subunit [p Ser293] Rabbit Polyclonal Antibody (FITC (Fluorescein))
Katalog # NOVUNB110-93479F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:pyruvate dehydrogenase (lipoamide) alpha 1
- Antigen symbol:PDHA1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:5160
- Antigen synonyms:PDHCE1A|PDHAD|PHE1A|PDHA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: Pyruvate Dehydrogenase E1-alpha subunit [p Ser293]
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat