Anti-BAI1 Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNB110-81586C
- Antikroppstyp:Primär
- Antikroppsnamn:Adhesion G Protein-Coupled Receptor B1
- Antigensymbol:ADGRB1
- Klonalitet:Polyclonal
- Konjugation:DyLight® 650
- Värd:Rabbit
- Isotyp:
- Reaktivitet:Human
- Environmentally Preferable:
- Gen-ID:575
- Antigen synonymer:GDAIF|BAI1|R75078|KIAA4089|Brain-Specific Angiogenesis Inhibitor 1
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:100 µL
- Förp.:0,1 mL
Type: Primary
Antigen: BAI1 (brain-specific angiogenesis inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse