Order Entry
Sweden
ContactUsLinkComponent
Anti-SLC9A8 Mouse Monoclonal Antibody (DyLight® 650) [clone: 7A11]
Anti-SLC9A8 Mouse Monoclonal Antibody (DyLight® 650) [clone: 7A11]
Catalog # NOVUNB110-62091C
Supplier:  Novus Biologicals
undefined
Anti-SLC9A8 Mouse Monoclonal Antibody (DyLight® 650) [clone: 7A11]
Catalog # NOVUNB110-62091C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sodium/hydrogen exchanger 8
  • Antigen symbol:
    NHE8
  • Clonality:
    Monoclonal
  • Clone:
    7A11
  • Conjugation:
    DyLight® 650
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    23315
  • Antigen synonyms:
    Sodium/hydrogen exchanger 8|Na (+)/H(+) exchanger 8|Solute carrier family 9 member 8|NHE-8|SLC9A8 antibody
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: SLC9A8 (solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8)
Clonality: monoclonal
Clone: 7A11
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat