Anti-DEFA5 Mouse monoclonal antibody unconjugated [clone: 8C8]
The Defensin alpha 5 Antibody (8C8) from Novus Biologicals is a mouse monoclonal antibody to Defensin alpha 5. This antibody reacts with human. The Defensin alpha 5 Antibody (8C8) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot.
Recommended Dilutions: Immunohistochemistry (Parafin): 5-10 ?g/ml
Type: Primary
Antigen: DEFA5
Clonality: Monoclonal
Clone: 8C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
- Catalog No:
- NOVUNB110-60002
- NOVUNB110-60002SS
- Antigen symbol:
- DEFA5
- DEFA5
- Antigen name:
- Defensin alpha 5
- Defensin alpha 5
- Antigen synonyms:
- HD-5|DEF5
- HD-5|DEF5
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 8C8
- 8C8
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 1670
- 1670
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice