Anti-KCNK2 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB110-41535V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:potassium channel, two pore domain subfamily K, member 2
- Antigen symbol:KCNK2
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- Western blot:Yes
- Gene ID:3776
- Antigen synonyms:hTREK-1e|hTREK-1c|TREK1|TPKC1|TREK-1|K2p2.1|TREK
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: KCNK2 (potassium channel, subfamily K, member 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: