Anti-Podocalyxin Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNB110-41503R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Podocalyxin
- Antigen symbol:PODX
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:5420
- Antigen synonyms:PCLP|PCLP-1|Gp200|PC
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Podocalyxin
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse