Anti-ATP13A2 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNB110-41486C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:ATPase type 13A2
- Antigen symbol:ATP13A2
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:23400
- Antigen synonyms:ATP13A2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ATP13A2 (ATPase type 13A2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse