Order Entry
Sweden
ContactUsLinkComponent
Anti-Aggrecan Neoepitope Rabbit Polyclonal Antibody
Anti-Aggrecan Neoepitope Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-Aggrecan Neoepitope Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-74350
  • NOVUNB100-74350SS
  • Antigen symbol:
  • Aggrecan Neoepitope
  • Aggrecan Neoepitope
  • Antigen name:
  • Aggrecan Neoepitope
  • Aggrecan Neoepitope
  • Antigen synonyms:
  • AGCAN|CSPG1|AGC1|SEDK|CSPGCP|MSK16
  • AGCAN|CSPG1|AGC1|SEDK|CSPGCP|MSK16
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 176
  • 176
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage buffer:
  • PBS and 30% Glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information