Anti-CAV1 Mouse Monoclonal Antibody [clone: 7C8]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: monoclonal
Clone: 7C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Specifications
- Catalog No:
- NOVUNB100-615
- NOVUNB100-615SS
- Antigen symbol:
- CAV1
- CAV1
- Antigen name:
- Caveolin 1
- Caveolin 1
- Antigen synonyms:
- CAV|VIP21|CAV1|MSTP085|caveolae protein 22kD|cell growth-inhibiting protein 32|caveolin 1
- CAV|VIP21|CAV1|MSTP085|caveolae protein 22kD|cell growth-inhibiting protein 32|caveolin 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 7C8
- 7C8
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 857
- 857
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice