Anti-NR4A1 Rabbit Polyclonal Antibody
NOVUNB100-56745
- Antikroppstyp:Primär
- Antikroppsnamn:Nuclear Receptor Subfamily 4 Group A Member 1
- Antigensymbol:NR4A1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Immunutfällning:Yes
- Reaktivitet:Human
- Western blot:Yes
- ChIP:Yes
- Gen-ID:3164
- Antigen synonymer:Orphan nuclear receptor HMR|HMR|ST-59|Orphan nuclear receptor NR4A1|GFRP1|NR4A1|NAK1|Early response protein NAK1|TR3 orphan receptor
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:50 µg
- Förp.:0,1 mg
Type: Primary
Antigen: NR4A1 (nuclear receptor subfamily 4, group A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse