Anti-Histone H2B, member S Rabbit Polyclonal Antibody
NOVUNB100-56347
- Antibody type:Primary
- Antigen name:H2B histone family, member S (pseudogene)
- Antigen symbol:H2BFS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:54145
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: Histone H2B, member S
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: