Anti-Caspase 8 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The Caspase-8 Antibody - (active / cleaved) from Novus Biologicals is a rabbit polyclonal antibody to Caspase-8. This antibody reacts with human, mouse, rat, gerbil. The Caspase-8 Antibody - (active / cleaved) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000
Type: Primary
Antigen: Caspase 8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Gerbil
Specifications
- Catalog No:
- NOVUNB100-56116SS
- NOVUNB100-56116
- Antigen symbol:
- CASP8
- CASP8
- Antigen name:
- caspase 8, apoptosis-related cysteine peptidase
- caspase 8, apoptosis-related cysteine peptidase
- Antigen synonyms:
- FLICE|MCH5|CAP4|ALPS2B|Casp-8|MACH
- FLICE|MCH5|CAP4|ALPS2B|Casp-8|MACH
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 841
- 841
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage buffer:
- Whole antisera
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice