Anti-PDK1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The PDK-1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PDK-1. This antibody reacts with human. The PDK-1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry/Immunofluorescence.
There is 1 scientific publication using this antibody.
Type: Primary
Antigen: PDK-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB100-2383SS
- NOVUNB100-2383
- Antigen symbol:
- PDPK1
- PDPK1
- Antigen name:
- 3-phosphoinositide dependent protein kinase 1
- 3-phosphoinositide dependent protein kinase 1
- Antigen synonyms:
- 3-phosphoinositide dependent protein kinase 1|Phosphoinositide-dependent kinase-1|PDK1|PDPK2|PDPK2P|PRO0461
- 3-phosphoinositide dependent protein kinase 1|Phosphoinositide-dependent kinase-1|PDK1|PDPK2|PDPK2P|PRO0461
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 5163
- 5163
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage buffer:
- Tris-Glycine and 0,15M NaCl
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice