Order Entry
Sweden
ContactUsLinkComponent
Anti-SREBF1 Rabbit Polyclonal Antibody (DyLight® 405)
Anti-SREBF1 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB100-2215V
Supplier:  Novus Biologicals
Anti-SREBF1 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB100-2215V
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sterol regulatory element binding transcription factor 1
  • Antigen symbol:
    SREBF1
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 405
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Gene ID:
    6720
  • Antigen synonyms:
    bHLHd1|SREBP1|SREBP-1c
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: SREBF1 (sterol regulatory element binding transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: