Anti-SREBF1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNB100-2215F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Sterol regulatory element binding transcription factor 1
- Antigen symbol:SREBF1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Western blot:Yes
- Gene ID:6720
- Antigen synonyms:bHLHd1|SREBP1|SREBP-1c
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SREBF1 (sterol regulatory element binding transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: