Order Entry
Sweden
ContactUsLinkComponent
Anti-OGG1 Rabbit Polyclonal Antibody
Anti-OGG1 Rabbit Polyclonal Antibody
Leverantör:  Novus Biologicals
Certifikat
Anti-OGG1 Rabbit Polyclonal Antibody
Leverantör:  Novus Biologicals

Specifikationer

  • Catalog No:
  • NOVUNB100-106
  • NOVUNB100-106SS
  • Antigen symbol:
  • OGG1
  • OGG1
  • Antigen name:
  • 8-Oxoguanine glycosylase
  • 8-Oxoguanine glycosylase
  • Antigen synonyms:
  • OGH1|HMMH|MUTM|HOGG1|CGL-2|CCP-X|CTLA1|CTSGL2|CSP-C
  • OGH1|HMMH|MUTM|HOGG1|CGL-2|CCP-X|CTLA1|CTSGL2|CSP-C
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 4968
  • 4968
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifikationer

Relaterad information