Anti-CD168 Rabbit Polyclonal Antibody
Catalog # BSBTPA1592
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Hyaluronan-mediated Motility
- Antigen symbol:CD168
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:RHAMM
- Amino acid number:706 - 724
- Sequence:KEGNTNCYRAPMECQESWK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human CD168 (706-724aa KEGNTNCYRAPMECQESWK).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Hyaluronan mediated motility receptor(HMMR) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: CD168
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human