Anti-ADAMTS4 Rabbit Polyclonal Antibody
BSBTPA1479
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase With Thrombospondin Type 1 Motif, 4
- Antigen symbol:ADAMTS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Aggrecanase 1|ADAM-TS 4|ADMP-1|A disintegrin and metalloproteinase with thrombospondin motifs 4|ADAM-TS4
- Amino acid number:820 - 837
- Sequence:RRAQILEILRRRPWAGRK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human ADAMTS4 (820-837aa RRAQILEILRRRPWAGRK), different from the related rat and mouse sequences by three amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for A disintegrin and metalloproteinase with thrombospondin motifs 4(ADAMTS4) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: ADAMTS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat