Anti-Cullin 4B Rabbit Polyclonal Antibody
Catalog # BSBTPA1233
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cullin 4B
- Antigen symbol:CUL4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Zebrafish,Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:Cullin-4B
- Amino acid number:895 - 913
- Sequence:DRDYMERDKENPNQYNYIA
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Cullin 4B (895-913aa DRDYMERDKENPNQYNYIA), identical to the related rat and mouse sequences
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Cullin-4B (CUL4B) detection. Tested with WB, IHC-P in Human;Mouse;Rat;Zebrafish.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Cullin 4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish