Order Entry
Sweden
ContactUsLinkComponent
Anti-PSGR Rabbit Polyclonal Antibody
Anti-PSGR Rabbit Polyclonal Antibody
Catalog # ABCAAB129883-100
Leverantör:  Abcam
undefined
Anti-PSGR Rabbit Polyclonal Antibody
Catalog # ABCAAB129883-100
Leverantör:  Abcam

Specifikationer

  • Antibody type:
    Primary
  • Antigen name:
    olfactory receptor, family 51, subfamily E, member 2
  • Antigen symbol:
    OR51E2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifikationer

Om denna produkt

Rabbit polyclonal to PSGR.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PSGR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human