Anti-FOXO1A+FOXO4/AFX+FOXO3A+Foxo6 (phospho T32) Rabbit Monoclonal Antibody [clone: EPR2314(2)]
ABCAAB128876-100
: Abcam
- Antibody type:Primary
- Antigen name:Forkhead Box O4
- Antigen symbol:FOXO4
- Clonality:Monoclonal
- Clone:EPR2314
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Seq:1286
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pack type:Vial
- Pk:100 µl
Rabbit monoclonal [EPR2314(2)] to FOXO1A (phospho T24) + FOXO4/AFX (phospho T28) + FOXO3A (phospho T32) + Foxo6 (phospho T26).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: FOXO1A+FOXO4/AFX+FOXO3A+Foxo6
Clonality: Monoclonal
Clone: EPR2314(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human