Anti-Pyruvate Dehydrogenase E1-alpha subunit Mouse Monoclonal Antibody [clone: 8D10E6]
Catalog # ABCAAB110334-100
Supplier: Abcam
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Pyruvate Dehydrogenase E1-alpha subunit
- Klonalitet:Monoclonal
- Klon:8D10E6
- Konjugation:Unconjugated
- Flödescytometri:Yes
- Värd:Mouse
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG1
- Western blot:Yes
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Type: Primary
Antigen: Pyruvate Dehydrogenase E1-alpha subunit
Clonality: monoclonal
Clone: 8D10E6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: