Anti-Pyruvate Dehydrogenase E1-alpha subunit Mouse Monoclonal Antibody [clone: 9H9AF5]
Catalog # ABCAAB110330-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Pyruvate Dehydrogenase E1-alpha subunit
- Clonality:Monoclonal
- Clone:9H9AF5
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Pyruvate Dehydrogenase E1-alpha subunit
Clonality: monoclonal
Clone: 9H9AF5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: