Order Entry
Sweden
ContactUsLinkComponent
Anti-Pyruvate Dehydrogenase E1-alpha subunit Rabbit Polyclonal Antibody
Anti-Pyruvate Dehydrogenase E1-alpha subunit Rabbit Polyclonal Antibody
Catalog # ABCAAB97352-50
Supplier:  Abcam
Anti-Pyruvate Dehydrogenase E1-alpha subunit Rabbit Polyclonal Antibody
Catalog # ABCAAB97352-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Pyruvate Dehydrogenase E1-alpha subunit
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    50 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Pyruvate Dehydrogenase E1-alpha subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat