Order Entry
Sweden
ContactUsLinkComponent
Anti-CD11b+CD11c Mouse Monoclonal Antibody [clone: OX42] (Biotin)
Anti-CD11b+CD11c Mouse Monoclonal Antibody [clone: OX42] (Biotin)
Catalog # ABCAAB33827-50
Supplier:  Abcam
undefined
Anti-CD11b+CD11c Mouse Monoclonal Antibody [clone: OX42] (Biotin)
Catalog # ABCAAB33827-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD11b/c equivalent
  • Clonality:
    Monoclonal
  • Clone:
    OX-42
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Rat
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD11b/c equivalent
Clonality: monoclonal
Clone: OX-42
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat