Anti-I kappaB- beta Rabbit Polyclonal Antibody
BIRBORB99168-100
: Biorbyt
- Antibody type:Primary
- Antigen name:I kappa B beta
- Antigen symbol:IKBB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Modification:phospho-Thr19
- Storage buffer:Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Molecular weight:37 kDa
- Storage temperature:Cool
- Concentration:20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- Shipping temperature:Ice
- Immunogen:A synthesized peptide derived from human I kappaB- beta around the phosphorylation site of Threonine 19
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1:500-1:2000
Type: Primary
Antigen: I kappaB- beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human