Anti-HSP90AA1 Mouse Monoclonal Antibody
Catalog # BIRBORB95871-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Heat shock protein 90kDa alpha (cytosolic), class A member 1
- Antigen symbol:HSP90AA1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Western blot:Yes
- Storage buffer:PBS, pH 7,2, 50% glycerol, 0,09% azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: HSP90AA1 (heat shock protein 90kDa alpha (cytosolic), class A member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: