Anti-MPST Rabbit Polyclonal Antibody
BIRBORB95692-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Mercaptopyruvate sulfurtransferase
- Antigen symbol:MPST
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:HEPES with 0,15M NaCl, 0,01% BSA, 0,03% sodium azide, and 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:50 µg
- Pk:50 µG
Immunochemistry: 1:400; Western Blot: 1:2000
Type: Primary
Antigen: MPST (mercaptopyruvate sulfurtransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat