Anti-MAPK15 Rabbit Polyclonal Antibody
BIRBORB95190-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 15
- Antigen symbol:MAPK15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage buffer:0,1 M Tris-glycine, pH 7, 10% Glycerol, 0,01% Thimerosal
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Immunofluorescence: 1:100-1:200; Immunofluorescence: 1:100-1:200; Immunochemistry: 10 ?g/ml; Western Blot: 1:500-1:3000
Type: Primary
Antigen: MAPK15 (mitogen-activated protein kinase 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human