Anti-APOB Rabbit polyclonal antibody unconjugated
BIRBORB95130-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Apolipoprotein B
- Antigen symbol:APOB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Format:heat inactivated serum
- Storage buffer:0,05% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Immunochemistry: 1:100
Type: Primary
Antigen: APOB (apolipoprotein B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: