Anti-SRSF3 Rabbit Polyclonal Antibody
Catalog # BIRBORB95129-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Serine/arginine-rich Splicing Factor 3
- Antigen symbol:SRSF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Storage buffer:PBS (without Mg2+, Ca2+), pH 7,4, 150 mM sodium chloride, 0,02% sodium azide, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ELISA: 1:5000; Immunofluorescence: 1:100-1:500; Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:1000
Type: Primary
Antigen: SRSF3 (serine/arginine-rich splicing factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: