Anti-MPP5 Rabbit Polyclonal Antibody
Catalog # BIRBORB89976-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Membrane Protein, Palmitoylated 5 (MAGUK p55 subfamily member 5)
- Antigen symbol:MPP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, pH 7, 40% Glycerol, 0,01% Thimerosal
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 10 ?g/ml; Western Blot: 1:500-1:3000
Type: Primary
Antigen: MPP5 (membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human