Anti-EREG Rabbit Polyclonal Antibody
BIRBORB88353-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Epiregulin
- Antigen symbol:EREG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:20 mM Tris-HCl, pH 8.0, 20 mg/ml BSA, 0,05% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:50 µg
- Pk:50 µG
Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: EREG (epiregulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human