Anti-PFKFB3 Rabbit Polyclonal Antibody
Catalog # BIRBORB77956-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
- Antigen symbol:PFKFB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Recombinant protein
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: 1:20000-1:80000; Western Blot: 1:1000-1:2000
Type: Primary
Antigen: PFKFB3 (6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human