Anti-Klebsiella aerogens K15 Mouse Monoclonal Antibody
Catalog # BIRBORB76688-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Klebsiella aerogens K15
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3
- Reactivity:Bacteria
- Storage buffer:Purified antibody containing PBS, 10mM sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:LPS membrane extract from K.aerogenes
- Purification:purified by Protein A chromatography
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Klebsiella aerogens K15
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Bacteria