Anti-hnRNPK/J Mouse Monoclonal Antibody
Catalog # BIRBORB76671-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:HNRNPK/J
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Western blot:Yes
- Storage buffer:PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:hnRNPs purified by oligo dc affinity chromatography
- Purification:purified by Protein A chromatography
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: hnRNPK/J
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: