Order Entry
Sweden
ContactUsLinkComponent
Anti-SCRAPPER Rabbit Polyclonal Antibody
Anti-SCRAPPER Rabbit Polyclonal Antibody
Catalog # BIRBORB74931-100
Supplier:  Biorbyt
undefined
Anti-SCRAPPER Rabbit Polyclonal Antibody
Catalog # BIRBORB74931-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SCRAPPER
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: SCRAPPER
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat