Anti-Karyopherin alpha 2 Rabbit Polyclonal Antibody
Catalog # BIRBORB73968-100
Leverantör: Biorbyt
Specifikationer
- Antibody type:Primary
- Antigen name:Karyopherin alpha 2
- Antigen symbol:KPNA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:0,71 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: Karyopherin alpha 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human