Order Entry
Sweden
ContactUsLinkComponent
Anti-CD69 Mouse Monoclonal Antibody
Anti-CD69 Mouse Monoclonal Antibody
Katalog # BIRBORB69242-100
Leverantör:  Biorbyt
undefined
Anti-CD69 Mouse Monoclonal Antibody
Katalog # BIRBORB69242-100
Leverantör:  Biorbyt

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    CD69 molecule
  • Antigensymbol:
    CD69
  • Klonalitet:
    Monoclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flödescytometri:
    Yes
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG1
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Mått:
    ascites fluid
  • Storage buffer:
    0,03% sodium azide.
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified recombinant fragment of human CD69 expressed in E. Coli.
  • Rening:
    purified by affinity chromatography
  • Storlek:
    100 µl
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000; Flow Cytometry: 1:200-1:400; ELISA: 1:10000

Type: Primary
Antigen: CD69 (CD69 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human