Anti-TLR4 Rabbit Polyclonal Antibody
BIRBORB67563-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Toll-like Receptor 4
- Antigen symbol:TLR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Storage buffer:PBS, 50% glycerol, 0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Developed against a synthetic peptide corresponding to amino acids 420-435 of human TLR4.
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1-3 ?g/ml; Immunochemistry: 1:50
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: