Anti-MAPK1/3 Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Western Blot: 1:5000; Immunochemistry: 1:50
Type: Primary
Antigen: MAPK1/3 (mitogen-activated protein kinase 1/3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Specifications
- Catalog No:
- BIRBORB67517-100
- BIRBORB67517-25
- Antigen symbol:
- MAPK1/3
- MAPK1/3
- Antigen name:
- Mitogen-activated protein kinase 1/3
- Mitogen-activated protein kinase 1/3
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- A 35 residue synthetic peptide, corresponding to Erk1 MAP kinase with the CGG spacer group added and the peptide coupled to KLH.
- A 35 residue synthetic peptide, corresponding to Erk1 MAP kinase with the CGG spacer group added and the peptide coupled to KLH.
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Storage buffer:
- PBS, 50% glycerol, 0,09% sodium azide
- PBS, 50% glycerol, 0,09% sodium azide
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice