Anti-SOS2 Rabbit Polyclonal Antibody
Catalog # BIRBORB67259-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:son of sevenless homolog 2 (Drosophila)
- Antigen symbol:SOS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage buffer:1 x PBS (pH 7,4), 40% Glycerol, 0,02% Thimerosal.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:C-terminal region of human SOS2
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:500-1:2000
Type: Primary
Antigen: SOS2 (son of sevenless homolog 2 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human