Order Entry
Sweden
ContactUsLinkComponent
Anti-MOCOS Rabbit Polyclonal Antibody
Anti-MOCOS Rabbit Polyclonal Antibody
Catalog # BIRBORB67220-100
Supplier:  Biorbyt
undefined
Anti-MOCOS Rabbit Polyclonal Antibody
Catalog # BIRBORB67220-100
Supplier:  Biorbyt

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    molybdenum cofactor sulfurase
  • Antigensymbol:
    MOCOS
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Storage buffer:
    1 x PBS (pH 7,4), 40% Glycerol, 0,02% Thimerosal.
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    N-terminal region of human MOCOS
  • Rening:
    purified by affinity chromatography
  • Storlek:
    100 µl
  • Förp.:
    100 µl

Specifications

About this item

Western Blot: 1:500-1:1000

Type: Primary
Antigen: MOCOS (molybdenum cofactor sulfurase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human