Order Entry
Sweden
ContactUsLinkComponent
 
Anti-HnRNP Q Rabbit Polyclonal Antibody
  BIRBORB137577-50
 :  Biorbyt
undefined
Anti-HnRNP Q Rabbit Polyclonal Antibody
  BIRBORB137577-50
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen symbol:
    HNRNPQ
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Storage buffer:
    PBS (without Mg2+, Ca2+), pH 7,4, 150 mM sodium chloride, 0,02% sodium azide, 50% glycerol
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    SYNCRIP/HnRNP Q antibody was raised against synthetic peptide from human hnRNP Q.
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

ELISA: 1:20000; Immunofluorescence: 1:100-1:500; Immunochemistry: 20 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: HnRNP Q
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat