Anti-ZBTB40 Rabbit Polyclonal Antibody
Catalog # BIRBORB137331-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:zinc finger and BTB domain containing 40
- Antigen symbol:ZBTB40
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, 0,02% sodium azide
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:ZBTB40 antibody was raised against a 19 amino acid peptide near the amino terminus of human ZBTB40.
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
- Immunochemistry: 5 μg/ml; Western Blot: 0.5-1 μg/ml
Type: Primary
Antigen: ZBTB40 (zinc finger and BTB domain containing 40)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human